Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022408661.1 | XP_022264369.1 | 3 | 947 | 115 | |
XM_022408662.1 | XP_022264370.1 | 2 | 592 | 115 | ||
XM_542793.6 | XP_542793.3 | 2 | 1321 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |