Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSLTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005633876.3 | XP_005633933.1 | 834 | 185 | ||
XM_005633877.3 | XP_005633934.1 | 1004 | 185 | |||
XM_014106431.2 | XP_013961906.1 | 1058 | 185 | |||
XM_022408277.1 | XP_022263985.1 | 1159 | 185 | |||
XM_022408278.1 | XP_022263986.1 | 1457 | 185 | |||
XM_022408279.1 | XP_022263987.1 | 757 | 185 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Canis familiaris |