Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618374.3 | XP_005618431.1 | 2 | 284 | 126 | |
XM_536200.6 | XP_536200.1 | 2 | 265 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |