Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEUROD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628693.3 | XP_005628750.1 | 2 | 1030 | 118 | |
XM_539504.6 | XP_539504.1 | 2 | 1160 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |