Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632856.3 | XP_005632913.1 | 4 - 5 | 923 | 134 | |
XM_022407023.1 | XP_022262731.1 | 4 - 5 | 923 | 134 | ||
XM_022407024.1 | XP_022262732.1 | 4 - 5 | 875 | 134 | ||
XM_022407025.1 | XP_022262733.1 | 4 - 5 | 923 | 134 | ||
XM_022407026.1 | XP_022262734.1 | 4 - 5 | 977 | 134 | ||
XM_022407027.1 | XP_022262735.1 | 3 - 4 | 802 | 134 | ||
XM_022407028.1 | XP_022262736.1 | 4 - 5 | 1017 | 134 | ||
XM_022407029.1 | XP_022262737.1 | 4 - 5 | 923 | 134 | ||
XM_022407030.1 | XP_022262738.1 | 4 - 5 | 923 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |