Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022427747.1 | XP_022283455.1 | 2 | 834 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |