Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628933.3 | XP_005628990.1 | 5 | 985 | 162 | |
XM_022427822.1 | XP_022283530.1 | 5 | 1081 | 162 | ||
XM_022427823.1 | XP_022283531.1 | 5 | 1036 | 162 | ||
XM_022427824.1 | XP_022283532.1 | 5 | 1026 | 162 | ||
XM_022427825.1 | XP_022283533.1 | 4 | 1093 | 162 | ||
XM_022427826.1 | XP_022283534.1 | 5 | 792 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |