Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014122379.2 | XP_013977854.1 | 3 | 436 | 85 | |
XM_014122380.2 | XP_013977855.1 | 3 | 3444 | 85 | ||
XM_014122381.2 | XP_013977856.1 | 3 | 288 | 85 | ||
XM_846987.4 | XP_852080.2 | 3 | 436 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |