Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536862.6 | XP_536862.2 | 1805 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris |