Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022412527.1 | XP_022268235.1 | 5 | 532 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |