Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEX3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618406.2 | XP_005618463.1 | 2 | 1026 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |