Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634257.3 | XP_005634314.1 | 5 | 1933 | 175 | |
XM_005634258.3 | XP_005634315.1 | 5 | 1306 | 175 | ||
XM_005634259.3 | XP_005634316.1 | 5 | 2547 | 175 | ||
XM_005634262.3 | XP_005634319.1 | 5 | 1229 | 175 | ||
XM_014106708.2 | XP_013962183.1 | 5 | 1225 | 175 | ||
XM_542715.5 | XP_542715.2 | 5 | 2287 | 175 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |