Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003431728.4 | XP_003431776.1 | 5 - 6 | 645 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |