Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627150.3 | XP_005627207.1 | 2 | 886 | 82 | |
XM_538835.6 | XP_538835.1 | 2 | 811 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |