Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627586.3 | XP_005627643.1 | 3 | 192 | 188 | |
XM_005627587.3 | XP_005627644.1 | 4 | 353 | 188 | ||
XM_022426311.1 | XP_022282019.1 | 3 | 216 | 188 | ||
XM_849202.5 | XP_854295.2 | 3 | 513 | 188 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |