Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002989.1 | NP_001002989.1 | 281 | 168 | ||
GenBank mRNA | AY266681.1 | - | 281 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |