Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENT5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022416211.1 | XP_022271919.1 | 3 | 878 | 113 | |
XM_022416212.1 | XP_022271920.1 | 2 | 804 | 113 | ||
XM_549103.5 | XP_549103.2 | 2 | 771 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |