Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625881.3 | XP_005625938.1 | 3 | 808 | 146 | |
XM_005625882.3 | XP_005625939.1 | 2 | 5598 | 146 | ||
XM_005625883.3 | XP_005625940.1 | 2 | 711 | 146 | ||
XM_022424438.1 | XP_022280146.1 | 4 | 3256 | 146 | ||
XM_022424439.1 | XP_022280147.1 | 4 | 5391 | 146 | ||
XM_022424440.1 | XP_022280148.1 | 2 | 914 | 146 | ||
XM_538374.6 | XP_538374.2 | 2 | 523 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |