Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022406941.1 | XP_022262649.1 | 11 - 12 | 1663 | 77 | |
XM_541984.3 | XP_541984.2 | 11 - 12 | 1416 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |