Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFBKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637002.3 | XP_005637059.1 | 2 | 574 | 97 | |
XM_022410992.1 | XP_022266700.1 | 4 | 1292 | 97 | ||
XM_022410993.1 | XP_022266701.1 | 3 | 752 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |