Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARBI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631285.3 | XP_005631342.1 | 3 | 1068 | 152 | |
XM_005631286.3 | XP_005631343.1 | 3 | 1242 | 152 | ||
XM_005631287.2 | XP_005631344.1 | 3 | 1026 | 152 | ||
XM_005631288.3 | XP_005631345.1 | 3 | 1094 | 152 | ||
XM_540753.5 | XP_540753.2 | 3 | 1078 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |