Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617652.3 | XP_005617709.1 | 4 - 5 | 782 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |