Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMUB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624403.3 | XP_005624460.1 | 1047 | 172 | ||
XM_005624405.3 | XP_005624462.1 | 947 | 172 | |||
XM_014116520.2 | XP_013971995.1 | 1495 | 172 | |||
XM_022422902.1 | XP_022278610.1 | 1332 | 172 | |||
XM_022422903.1 | XP_022278611.1 | 1588 | 172 | |||
XM_022422904.1 | XP_022278612.1 | 1610 | 172 | |||
XM_022422905.1 | XP_022278613.1 | 905 | 172 | |||
XM_843644.5 | XP_848737.2 | 1063 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |