Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637593.3 | XP_005637650.1 | 6 | 613 | 136 | |
XM_005637596.3 | XP_005637653.1 | 4 | 423 | 136 | ||
XM_014108995.2 | XP_013964470.1 | 6 | 639 | 136 | ||
XM_014108996.2 | XP_013964471.1 | 6 | 684 | 136 | ||
XM_014108997.2 | XP_013964472.1 | 7 | 722 | 136 | ||
XM_014108999.2 | XP_013964474.1 | 5 | 546 | 136 | ||
XM_022411644.1 | XP_022267352.1 | 4 | 685 | 136 | ||
XM_022411645.1 | XP_022267353.1 | 7 | 756 | 136 | ||
XM_022411646.1 | XP_022267354.1 | 7 | 801 | 136 | ||
XM_022411647.1 | XP_022267355.1 | 7 | 696 | 136 | ||
XM_022411648.1 | XP_022267356.1 | 6 | 658 | 136 | ||
XM_534983.5 | XP_534983.2 | 5 | 520 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |