Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005642025.3 | XP_005642082.1 | 3 | 489 | 81 | |
XM_005642026.3 | XP_005642083.1 | 3 | 489 | 81 | ||
XM_005642027.3 | XP_005642084.1 | 3 | 487 | 81 | ||
XM_005642028.3 | XP_005642085.1 | 3 | 488 | 81 | ||
XM_848512.4 | XP_853605.1 | 3 | 486 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |