Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_847542.5 | XP_852635.1 | 1126 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris |