Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014106423.2 | XP_013961898.1 | 2 - 3 | 357 | 104 | |
XM_022408029.1 | XP_022263737.1 | 2 - 3 | 290 | 104 | ||
XM_534146.6 | XP_534146.4 | 3 - 4 | 317 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |