Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_546754.5 | XP_546754.2 | 1086 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris |