Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_546751.5 | XP_546751.2 | 2319 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |