Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_847757.3 | XP_852850.1 | 7 - 8 | 623 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |