Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022422428.1 | XP_022278136.1 | 3 - 4 | 314 | 195 | |
XM_022422431.1 | XP_022278139.1 | 3 - 4 | 343 | 195 | ||
XM_022422432.1 | XP_022278140.1 | 3 - 4 | 342 | 195 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |