Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_860793.5 | XP_865886.3 | 1 - 2 | 172 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |