Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048130.1 | NP_001041595.1 | 5 - 6 | 557 | 143 | |
XM_005638180.3 | XP_005638237.1 | 6 - 7 | 713 | 143 | ||
XM_005638181.2 | XP_005638238.1 | 6 - 7 | 737 | 143 | ||
XM_022412499.1 | XP_022268207.1 | 5 - 6 | 510 | 143 | ||
GenBank mRNA | DQ494380.1 | - | 5 - 6 | 557 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |