Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417955.1 | XP_022273663.1 | 3 | 200 | 143 | |
XM_022417956.1 | XP_022273664.1 | 3 | 191 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |