Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003435229.4 | XP_003435277.3 | 264 | 152 | ||
XM_005624449.3 | XP_005624506.2 | 430 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |