Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616083.3 | XP_005616140.2 | 9 | 4862 | 177 | |
XM_014114388.2 | XP_013969863.1 | 10 | 5105 | 177 | ||
XM_014114392.2 | XP_013969867.1 | 12 | 5270 | 177 | ||
XM_022422507.1 | XP_022278215.1 | 10 | 4878 | 177 | ||
XM_022422508.1 | XP_022278216.1 | 12 | 5043 | 177 | ||
XM_022422509.1 | XP_022278217.1 | 9 | 4631 | 177 | ||
GenBank mRNA | HM771320.1 | - | 1 | 2648 | 177 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |