Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256018.1 | NP_001242947.1 | 2 | 124 | 90 | |
NM_001256262.1 | NP_001243191.1 | 2 | 168 | 90 | ||
XM_014114980.2 | XP_013970455.1 | 2 | 265 | 90 | ||
XM_022420763.1 | XP_022276471.1 | 2 | 213 | 90 | ||
XM_022420766.1 | XP_022276474.1 | 3 | 255 | 90 | ||
GenBank mRNA | DN338485.1 | - | 2 | 124 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |