Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014375.1 | NP_001014397.1 | 4 | 1053 | 59 | |
XM_005627064.3 | XP_005627121.1 | 4 | 1116 | 59 | ||
XM_005627065.3 | XP_005627122.1 | 4 | 1184 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |