Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003074.1 | NP_001003074.1 | 3 - 4 | 2044 | 106 | |
GenBank mRNA | AF177470.1 | - | 3 - 4 | 2044 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |