Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005264.3 | NP_001005264.2 | 1 | 2221 | 110 | |
XM_005628829.3 | XP_005628886.1 | 2 | 2266 | 110 | ||
XM_022427656.1 | XP_022283364.1 | 2 | 2316 | 110 | ||
XM_022427657.1 | XP_022283365.1 | 2 | 2294 | 110 | ||
GenBank mRNA | AB189639.1 | - | 1 | 2271 | 110 | |
EU487534.1 | - | 1 | 2221 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |