Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048117.1 | NP_001041582.1 | 713 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |