Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097551.1 | NP_001091020.1 | 5 - 6 | 434 | 77 | |
XM_022426505.1 | XP_022282213.1 | 5 - 6 | 439 | 77 | ||
GenBank mRNA | EF121758.1 | - | 5 - 6 | 434 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |