Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097551.1 | NP_001091020.1 | 9 - 10 | 822 | 121 | |
GenBank mRNA | EF121758.1 | - | 9 - 10 | 822 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |