Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097557.1 | NP_001091026.1 | 8 - 9 | 1482 | 116 | |
XM_005615775.2 | XP_005615832.1 | 9 - 10 | 1662 | 116 | ||
GenBank mRNA | DQ975208.1 | - | 8 - 9 | 1482 | 116 | |
DQ975210.1 | - | 6 - 7 | 948 | 116 | ||
DQ975212.1 | - | 5 - 6 | 672 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |