Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109958.1 | NP_001103428.1 | 3 - 4 | 420 | 88 | |
GenBank mRNA | AM748733.1 | - | 3 - 4 | 420 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |