Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109960.1 | NP_001103430.1 | 5 - 6 | 378 | 68 | |
XM_014119221.2 | XP_013974696.1 | 5 - 6 | 418 | 68 | ||
GenBank mRNA | AB299408.1 | - | 5 - 6 | 378 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |