Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_850342.4 | XP_855435.1 | 1 - 2 | 512 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |