Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130830.1 | NP_001124302.1 | 4 - 5 | 894 | 82 | |
GenBank mRNA | EU373510.1 | - | 4 - 5 | 894 | 82 | |
KT361522.1 | - | 4 - 5 | 896 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |