Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142257.1 | NP_001135729.1 | 6 - 7 | 617 | 67 | |
NM_001308549.1 | NP_001295478.1 | 8 - 9 | 732 | 67 | ||
XM_005632296.3 | XP_005632353.1 | 6 - 7 | 563 | 67 | ||
XM_005632301.3 | XP_005632358.1 | 5 - 6 | 500 | 67 | ||
XM_014121714.2 | XP_013977189.1 | 7 - 8 | 699 | 67 | ||
XM_014121717.2 | XP_013977192.1 | 6 - 7 | 602 | 67 | ||
XM_014121719.2 | XP_013977194.1 | 5 - 6 | 520 | 67 | ||
XM_022406193.1 | XP_022261901.1 | 9 - 10 | 823 | 67 | ||
XM_022406195.1 | XP_022261903.1 | 9 - 10 | 827 | 67 | ||
XM_022406196.1 | XP_022261904.1 | 7 - 8 | 643 | 67 | ||
XM_022406197.1 | XP_022261905.1 | 8 - 9 | 698 | 67 | ||
XM_022406198.1 | XP_022261906.1 | 6 - 7 | 636 | 67 | ||
XM_022406199.1 | XP_022261907.1 | 5 - 6 | 498 | 67 | ||
XM_022406200.1 | XP_022261908.1 | 5 - 6 | 497 | 67 | ||
GenBank mRNA | AB469369.1 | - | 6 - 7 | 606 | 67 | |
AB469370.1 | - | 6 - 7 | 617 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |