Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164486.1 | NP_001157958.1 | 5 - 6 | 690 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |